Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Chlorobenzyl chloride
CAS 620-20-2
Molecular Formula C7H6Cl2
Formula Weight 161.03
MDL Number MFCD00000905
UN Number UN2235
Beilstein 742266
Synonym alpha,3-Dichlorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1104114

 
alfa aesar™
A1104114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.03
Chemical Name or Material 3-Chlorobenzyl chloride
TSCA TSCA
Odor Pungent
Assay 97%
Molecular Formula C7H6Cl2
MDL Number MFCD00000905
CAS 620-20-2
UN Number UN2235
Beilstein 742266
Synonym alpha,3-Dichlorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROBENZYL CHLORIDES, LIQUID
Flash Point 98°C (208°F)
Boiling Point 215° to 216°C
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.