Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzyl alcohol, 99%

$66.50 - $1,019.00

Chemical Identifiers

Chemical Name or Material 3-Chlorobenzyl alcohol
CAS 873-63-2
Molecular Formula C7H7ClO
Formula Weight 142.59
MDL Number MFCD00004632
Beilstein 2041501
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1184109

  SDS
alfa aesar™
A1184109
10g Each for $66.50

AAA1184130

  SDS
alfa aesar™
A1184130
250g Each for $1,019.00

AAA1184118

  SDS
alfa aesar™
A1184118
50g Each for $261.68
Description & Specifications

Specifications

Formula Weight 142.59
Chemical Name or Material 3-Chlorobenzyl alcohol
Assay 99%
Molecular Formula C7H7ClO
MDL Number MFCD00004632
CAS 873-63-2
Beilstein 2041501
Flash Point >110°C (230°F)
Boiling Point 236° to 237°C
Quantity 10g