Please login to your online account to display your discounted pricing

3-Chlorobenzoyl Chloride 99+%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  108745000

 View more versions of this product

Catalog No. AC108745000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 175.01
Physical Form Liquid
Assay ≥99.0% (acid halide), carboxylic acid (as chlorobenzoic acid): <1.0%
Quantity 500mL
Color Clear colorless
Beilstein 09, 338
TSCA TSCA
EINECS Number 210-552-6
CAS Max % 100.0
CAS Min % 99.0
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99+%
Name Note 99+%
CAS 618-46-2
Chemical Name or Material 3-Chlorobenzoyl chloride
MDL Number MFCD00000671
Molecular Formula C7H4Cl2O
Linear Formula ClC6H4COCl
Refractive Index 1.5680 to 1.5700 (20°C, 589nm)
Packaging Glass bottle
Impurities total other poly-halogenated acid halides: < 0.03%
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material 3-Chlorobenzoyl chloride
Name Note 99+%
CAS 618-46-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0% (acid halide), carboxylic acid (as chlorobenzoic acid): <1.0%
Assay Percent Range 99+%
Molecular Formula C7H4Cl2O
Linear Formula ClC6H4COCl
Formula Weight 175.01
MDL Number MFCD00000671
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents