missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108740500

 View more versions of this product

Catalog No. AC108740500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chlorobenzoyl chloride
618-46-2
100.0
1.3600g/mL
110°C
0.03% max. Total other poly-halogenated acid halides (GC)
C7H4Cl2O
ClC6H4COCl
50 mL
1.36
Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and acetone
ClC(=O)C1=CC=CC(Cl)=C1
175.01
175.01
Liquid
99+%
99.0
Colorless
223.0°C to 225.0°C
Authentic
Glass bottle
1.5680 to 1.5700
MFCD00000671
09, 338
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
WHIHIKVIWVIIER-UHFFFAOYSA-N
3-chlorobenzoyl chloride
69252
99+%

Chemical Identifiers

618-46-2
175.01
WHIHIKVIWVIIER-UHFFFAOYSA-N
69252
ClC(=O)C1=CC=CC(Cl)=C1
C7H4Cl2O
MFCD00000671
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
3-chlorobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-552-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only