missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzotrifluoride, TCI America™

$48.89 - $400.28

Chemical Identifiers

CAS 98-15-7
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000594
InChI Key YTCGOUNVIAWCMG-UHFFFAOYSA-N
Synonym 3-chlorobenzotrifluoride, 1-chloro-3-trifluoromethyl benzene, m-chlorobenzotrifluoride, benzene, 1-chloro-3-trifluoromethyl, 3-chloro-alpha,alpha,alpha-trifluorotoluene, 3-chlorotrifluoromethylbenzene, m-trifluoromethylphenyl chloride, meta trifluoromethyl chlorobenzene, 3-chlorotrifluorotoluene, ccris 2810
PubChem CID 7374
IUPAC Name 1-chloro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)Cl)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C030425G View Documents TCI America
C030425G
25g Each for $48.89
 
C0304500G View Documents TCI America
C0304500G
500g Each for $400.28
 
Specifications

Chemical Identifiers

98-15-7
180.554
YTCGOUNVIAWCMG-UHFFFAOYSA-N
7374
C1=CC(=CC(=C1)Cl)C(F)(F)F
C7H4ClF3
MFCD00000594
3-chlorobenzotrifluoride, 1-chloro-3-trifluoromethyl benzene, m-chlorobenzotrifluoride, benzene, 1-chloro-3-trifluoromethyl, 3-chloro-alpha,alpha,alpha-trifluorotoluene, 3-chlorotrifluoromethylbenzene, m-trifluoromethylphenyl chloride, meta trifluoromethyl chlorobenzene, 3-chlorotrifluorotoluene, ccris 2810
1-chloro-3-(trifluoromethyl)benzene

Specifications

139°C
25g
C7H4ClF3
2234
YTCGOUNVIAWCMG-UHFFFAOYSA-N
1-chloro-3-(trifluoromethyl)benzene
7374
Clear Liquid at 20°C
3-Chlorobenzotrifluoride
Yellow
98-15-7
MFCD00000594
3-chlorobenzotrifluoride, 1-chloro-3-trifluoromethyl benzene, m-chlorobenzotrifluoride, benzene, 1-chloro-3-trifluoromethyl, 3-chloro-alpha,alpha,alpha-trifluorotoluene, 3-chlorotrifluoromethylbenzene, m-trifluoromethylphenyl chloride, meta trifluoromethyl chlorobenzene, 3-chlorotrifluorotoluene, ccris 2810
C1=CC(=CC(=C1)Cl)C(F)(F)F
180.554
180.55
≥90.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0053

RTECSNumber : XS9142000

TSCA : Yes