Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzotrifluoride, 98%

Chemical Identifiers

Chemical Name or Material 3-Chlorobenzotrifluoride
CAS 98-15-7
Molecular Formula C7H4ClF3
Formula Weight 180.56
MDL Number MFCD00000594
UN Number UN2234
Beilstein 510215
Synonym 3-Chloro-alpha,alpha,alpha-trifluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1985422

 
alfa aesar™
A1985422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1985414

 
alfa aesar™
A1985414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.56
Chemical Name or Material 3-Chlorobenzotrifluoride
TSCA TSCA
Assay 98%
Molecular Formula C7H4ClF3
MDL Number MFCD00000594
CAS 98-15-7
UN Number UN2234
Beilstein 510215
Synonym 3-Chloro-alpha,alpha,alpha-trifluorotoluene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES
RTECS Number XS9142000
Flash Point 38°C (100°F)
Boiling Point 137° to 138°C
Melting Point −56°C
Quantity 100g

Applications
3-Chlorobenzotrifluoride is used as an intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water <0.1 g/100 mL at 22°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.