missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chlorobenzonitrile, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC108680500

 View more versions of this product

Catalog No. AC108680500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5% (GC)
38° to 42°C
50g
1.1400g/mL
100.0
766-84-7
C7H4ClN
3-Chlorobenzonitrile
MFCD00001798
137.57
Liquid
Clear colorless to yellow
09, 339
Glass bottle
Authentic
ClC6H4CN
99%
98.5

Chemical Identifiers

766-84-7
100.0
C7H4ClN
137.57
3-Chlorobenzonitrile
98.5
99%
ClC6H4CN
MFCD00001798
≥98.5% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning