missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzoic hydrazide +98%, ACROS Organics™

Chemical Identifiers

CAS 1673-47-8
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C7H7ClN2O
Linear Formula ClC6H4C(=O)NHNH2
Formula Weight 170.6
MDL Number MFCD00014757
Chemical Name or Material 3-Chlorobenzoic hydrazide
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC393200250 SDS Thermo Fisher Scientific
AC393200250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1673-47-8
100.0
C7H7ClN2O
170.6
3-Chlorobenzoic hydrazide
98.0
98+%
ClC6H4C(=O)NHNH2
MFCD00014757
≥98.0%

Specifications

Crystalline Powder
25g
100.0
Glass bottle
1673-47-8
MFCD00014757
98+%
≥98.0%
170.6
White
98.0
Authentic
C7H7ClN2O
ClC6H4C(=O)NHNH2
3-Chlorobenzoic hydrazide

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.