Please login to your online account to display your discounted pricing

3-Chlorobenzoic acid, 99+%, ACROS Organics™

$31.99 - $92.87

Chemical Identifiers

Chemical Name or Material 3-Chlorobenzoic acid
CAS 535-80-8
CAS Min % 99.0
CAS Max % 100.0
Assay 99.0 to 100.5%
Assay Percent Range 99+%
Molecular Formula C7H5ClO2
Linear Formula ClC6H4CO2H
Formula Weight 156.57
MDL Number MFCD00002491
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC108665000

  SDS
thermo fisher scientific
AC108665000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC108660250

  SDS
fisher scientific acros
AC108660250
Plastic bottle 25g Each for $31.99

AC108661000

  SDS
fisher scientific acros
108661000
Plastic bottle 100g Each for $92.87
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 156.57
Quantity 500g
Color White to beige
Assay 99.0 to 100.5%
Melting Point 154° to 157°C
Beilstein 09, 337
Merck Index 15, 2124
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 3-Chlorobenzoic acid
Assay Percent Range 99+%
Molecular Formula C7H5ClO2
CAS 535-80-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00002491
Linear Formula ClC6H4CO2H
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Plastic bottle
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.