missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzoic acid, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108660250

 View more versions of this product

Catalog No. AC108660250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chlorobenzoic acid
99.0
154.0°C to 157.0°C
150°C
99% min. (HPLC)
C7H5ClO2
MFCD00002491
09, 337
m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
LULAYUGMBFYYEX-UHFFFAOYSA-N
3-chlorobenzoic acid
447
156.57
Crystalline Powder
535-80-8
100.0
White to Beige
Authentic
Plastic bottle
ClC6H4CO2H
25 g
15, 2124
Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
C1=CC(=CC(=C1)Cl)C(=O)O
156.57
CHEBI:49410
99+%

Chemical Identifiers

535-80-8
LULAYUGMBFYYEX-UHFFFAOYSA-N
447
3-chlorobenzoic acid
MFCD00002491
m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
CHEBI:49410
C1=CC(=CC(=C1)Cl)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-618-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only