Please login to your online account to display your discounted pricing

3-Chlorobenzoic acid, 99+%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  108661000

 View more versions of this product

Catalog No. AC108661000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 156.57
Quantity 100g
Color White to beige
Beilstein 09, 337
Merck Index 15, 2124
Assay 99.0 to 100.5%
Melting Point 154° to 157°C
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 3-Chlorobenzoic acid
Assay Percent Range 99+%
Molecular Formula C7H5ClO2
CAS 535-80-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula ClC6H4CO2H
MDL Number MFCD00002491
Health Hazard 1 Warning
Infrared Spectrum Authentic
Packaging Plastic bottle

Specifications

Chemical Name or Material 3-Chlorobenzoic acid
CAS 535-80-8
CAS Min % 99.0
CAS Max % 100.0
Assay 99.0 to 100.5%
Assay Percent Range 99+%
Molecular Formula C7H5ClO2
Linear Formula ClC6H4CO2H
Formula Weight 156.57
MDL Number MFCD00002491
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents