Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzo[b]thiophene-2-carboxamide, 98%

$64.18 - $173.28

Chemical Identifiers

Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carboxamide
CAS 21211-09-6
Molecular Formula C9H6ClNOS
Formula Weight 211.67
MDL Number MFCD00067793
Beilstein 1425601
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1889803

  SDS
alfa aesar™
L1889803
1g Each for $173.28

AAL18898MD

  SDS
alfa aesar™
L18898MD
250mg Each for $64.18
Description & Specifications

Specifications

Formula Weight 211.67
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carboxamide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C9H6ClNOS
MDL Number MFCD00067793
Beilstein 1425601
Melting Point 234° to 236°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 21211-09-6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.