Please login to your online account to display your discounted pricing

3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 97%, Maybridge™


Quantity 10g

Manufacturer: fisher scientific company  BTB00300EA

 View more versions of this product

Catalog No. BTB00300EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 231.1
Quantity 10g
MDL Number MFCD00053069
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 21815-91-8
Molecular Formula C9H4Cl2OS
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carbonyl chloride

Specifications

Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carbonyl chloride
Assay Percent Range 97%
Molecular Formula C9H4Cl2OS
Formula Weight 231.1
MDL Number MFCD00053069