Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzhydrazide, 98+%

$139.07 - $569.55

Chemical Identifiers

Chemical Name or Material 3-Chlorobenzhydrazide
CAS 1673-47-8
Molecular Formula C7H7ClN2O
Formula Weight 170.6
MDL Number MFCD00014757
Beilstein 973243
Synonym 3-Chlorobenzoic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0107109

  SDS
alfa aesar™
L0107109
10g Each for $139.07

AAL0107118

  SDS
alfa aesar™
L0107118
50g Each for $569.55
Description & Specifications

Specifications

Formula Weight 170.6
Chemical Name or Material 3-Chlorobenzhydrazide
CAS 1673-47-8
MDL Number MFCD00014757
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3-Chlorobenzoic acid hydrazide
Health Hazard 1 Warning
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 156° to 160°C
Beilstein 973243
Quantity 10g
Molecular Formula C7H7ClN2O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.