missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1028306

 View more versions of this product

Catalog No. AAA1028306



Specifications

Specifications

3-Chlorobenzenesulfonyl chloride
102°C to 104°C (1mmHg)
5g
C11H14O
UN3265
3-methyl-1-phenyl-2-butanone, benzyl isopropyl ketone, isopropylbenzyl ketone, 2-butanone, 3-methyl-1-phenyl, 1-phenyl-3-methyl-2-butanone, unii-81804s3mkm, isopropyl benzyl ketone, isopropylbenzylketon, 5-nitrophenyl laurate, 2-methyl-4-phenyl-3-butanone
CC(C)C(=O)CC1=CC=CC=C1
162.23
211.07
>110°C (230°F)
~21°C
2893-05-2
MFCD00051697
2937139
ZATIMOAGKJEVGN-UHFFFAOYSA-N
3-methyl-1-phenylbutan-2-one
76163
98%

Chemical Identifiers

2893-05-2
162.23
ZATIMOAGKJEVGN-UHFFFAOYSA-N
76163
CC(C)C(=O)CC1=CC=CC=C1
C11H14O
MFCD00051697
3-methyl-1-phenyl-2-butanone, benzyl isopropyl ketone, isopropylbenzyl ketone, 2-butanone, 3-methyl-1-phenyl, 1-phenyl-3-methyl-2-butanone, unii-81804s3mkm, isopropyl benzyl ketone, isopropylbenzylketon, 5-nitrophenyl laurate, 2-methyl-4-phenyl-3-butanone
3-methyl-1-phenylbutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents