missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chlorobenzenesulfonamide, 98%

Manufacturer:  Alfa Aesar™ L0834906

 View more versions of this product

Catalog No. AAL0834906



Description

Description

Applications
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Chlorobenzenesulfonamide
5g
C6H6ClNO2S
2832245
WSYQJNPRQUFCGL-UHFFFAOYSA-N
3-chlorobenzenesulfonamide
519377
98%
144°C to 148°C
17260-71-8
MFCD00051976
3-chloranylbenzenesulfonamide
C1=CC(=CC(=C1)Cl)S(=O)(=O)N
191.629
191.64

Chemical Identifiers

17260-71-8
191.629
WSYQJNPRQUFCGL-UHFFFAOYSA-N
519377
C1=CC(=CC(=C1)Cl)S(=O)(=O)N
C6H6ClNO2S
MFCD00051976
3-chloranylbenzenesulfonamide
3-chlorobenzenesulfonamide
Documents