missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chlorobenzaldehyde, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC108615000

 View more versions of this product

Catalog No. AC108615000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-chlorobenzaldehyde
Clear colorless to light yellow
587-04-2
100.0
Glass bottle
Liquid
99%
ClC6H4CHO
MFCD00003350
1.241g/mL
500mL
98.5
Authentic
1.5630 to 1.5650 (20°C, 589nm)
≥98.5%
C7H5ClO
140.57
07, 234

Chemical Identifiers

587-04-2
100.0
C7H5ClO
140.57
3-chlorobenzaldehyde
98.5
99%
ClC6H4CHO
MFCD00003350
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.