missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloroaniline Hydrochloride, TCI America™

$55.00 - $252.09

Chemical Identifiers

CAS 141-85-5
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00035485
InChI Key NMGHWHCTRGZZOP-UHFFFAOYSA-N
Synonym 3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
PubChem CID 67336
IUPAC Name 3-chloroaniline;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C011325G View Documents TCI America
C011325G
25g Each for $55.00
 
C0113500G View Documents TCI America
C0113500G
500g Each for $252.09
 
Specifications

Chemical Identifiers

141-85-5
164.029
NMGHWHCTRGZZOP-UHFFFAOYSA-N
67336
C1=CC(=CC(=C1)Cl)N.Cl
C6H7Cl2N
MFCD00035485
3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
3-chloroaniline;hydrochloride

Specifications

Undesignated
141-85-5
MFCD00035485
NMGHWHCTRGZZOP-UHFFFAOYSA-N
3-chloroaniline;hydrochloride
67336
Crystal-Powder at 20°C
3-Chloroaniline Hydrochloride
25g
C6H7Cl2N
3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
C1=CC(=CC(=C1)Cl)N.Cl
164.029
164.03
≥99.0% (HPLC,N)
Safety and Handling

Safety and Handling

RTECSNumber : BX1393800

TSCA : Yes