Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloroaniline hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloroaniline hydrochloride
CAS 141-85-5
Molecular Formula C6H6ClN·HCl
Formula Weight 164.03
MDL Number MFCD00035485
UN Number UN2018
Beilstein 3623168
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1348414

  SDS
alfa aesar™
L1348414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1348422

  SDS
alfa aesar™
L1348422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.03
Chemical Name or Material 3-Chloroaniline hydrochloride
CAS 141-85-5
Molecular Formula C6H6ClN·HCl
MDL Number MFCD00035485
Health Hazard 1 Danger
UN Number UN2018
Health Hazard 3 GHS P Statement
P280h-P273-P309-P310-P302+P352-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Characteristic
Melting Point ≈222°C
Assay 97%
Quantity 25g
TSCA TSCA
Beilstein 3623168
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
RTECS Number BX1393800
Health Hazard 2 GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Safety and Handling

Danger

GHS P Statement
P280h-P273-P309-P310-P302+P352-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.