missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloroaniline hydrochloride, 97%

Manufacturer:  Alfa Aesar™ L1348422

 View more versions of this product

Catalog No. AAL1348422



Specifications

Specifications

3-Chloroaniline hydrochloride
~222°C
141-85-5
MFCD00035485
3623168
NMGHWHCTRGZZOP-UHFFFAOYSA-N
3-chloroaniline;hydrochloride
67336
97%
Characteristic
100g
C6H7Cl2N
UN2018
3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
C1=CC(=CC(=C1)Cl)N.Cl
164.029
164.03

Chemical Identifiers

141-85-5
164.029
NMGHWHCTRGZZOP-UHFFFAOYSA-N
67336
C1=CC(=CC(=C1)Cl)N.Cl
C6H7Cl2N
MFCD00035485
3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
3-chloroaniline;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P280h-P273-P309-P310-P302+P352-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID

RTECSNumber : BX1393800

TSCA : TSCA

Documents