missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloroacetophenone, 98+%

Manufacturer:  Alfa Aesar™ A1559206

 View more versions of this product

Catalog No. AAA1559206



Specifications

Specifications

3'-Chloroacetophenone
227°C to 229°C
99-02-5
MFCD00000593
636318
UUWJBXKHMMQDED-UHFFFAOYSA-N
1-(3-chlorophenyl)ethanone
14933
≥98%
105°C (221°F)
5g
C8H7ClO
UN3416
3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
CC(=O)C1=CC(=CC=C1)Cl
154.593
154.6

Chemical Identifiers

99-02-5
154.593
UUWJBXKHMMQDED-UHFFFAOYSA-N
14933
CC(=O)C1=CC(=CC=C1)Cl
C8H7ClO
MFCD00000593
3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
1-(3-chlorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID

TSCA : TSCA

Documents