missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloroacetanilide, 98+%

Manufacturer:  Alfa Aesar™ A1492114

 View more versions of this product

Catalog No. AAA1492114



Specifications

Specifications

3'-Chloroacetanilide
25g
C8H8ClNO
2208105
3'-chloroacetanilide, 3-chloroacetanilide, n-3-chlorophenyl acetamide, m-chloroacetanilide, acetamide, n-3-chlorophenyl, acetanilide, 3'-chloro, meta-chloroacetanilide, unii-74h09hu61j, acetic acid, amide, n 3-chlorophenyl, m-chloroacetoanilide
CC(=O)NC1=CC(=CC=C1)Cl
169.608
169.61
75°C to 79°C
588-07-8
MFCD00016341
14,2111
MUUQHCOAOLLHIL-UHFFFAOYSA-N
N-(3-chlorophenyl)acetamide
11493
≥98%

Chemical Identifiers

588-07-8
169.608
MUUQHCOAOLLHIL-UHFFFAOYSA-N
11493
CC(=O)NC1=CC(=CC=C1)Cl
C8H8ClNO
MFCD00016341
3'-chloroacetanilide, 3-chloroacetanilide, n-3-chlorophenyl acetamide, m-chloroacetanilide, acetamide, n-3-chlorophenyl, acetanilide, 3'-chloro, meta-chloroacetanilide, unii-74h09hu61j, acetic acid, amide, n 3-chlorophenyl, m-chloroacetoanilide
N-(3-chlorophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AE1000100

TSCA : TSCA

Documents