Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloroacetanilide, 98+%

$33.76 - $339.03

Chemical Identifiers

Chemical Name or Material 3'-Chloroacetanilide
CAS 588-07-8
Molecular Formula C8H8ClNO
Formula Weight 169.61
MDL Number MFCD00016341
Beilstein 2208105
Merck Index 14,2111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1492114

  SDS
alfa aesar™
A1492114
25g Each for $33.76

AAA1492122

  SDS
alfa aesar™
A1492122
100g Each for $84.82

AAA1492136

  SDS
alfa aesar™
A1492136
500g Each for $339.03
Description & Specifications

Specifications

Formula Weight 169.61
Chemical Name or Material 3'-Chloroacetanilide
TSCA TSCA
Assay ≥98%
Molecular Formula C8H8ClNO
MDL Number MFCD00016341
CAS 588-07-8
Beilstein 2208105
Merck Index 14,2111
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AE1000100
Melting Point 75° to 79°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.