missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-p-cresol, TCI America™

Manufacturer:  TCI America C333325G

 View more versions of this product

Catalog No. C333325G



Specifications

Specifications

3-Chloro-p-cresol
Undesignated
25g
C7H7ClO
3437
VQZRLBWPEHFGCD-UHFFFAOYSA-N
3-chloro-4-methylphenol
14853
Crystal-Lump at 20°C
228°C
53°C
615-62-3
MFCD00060319
phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol
CC1=C(C=C(C=C1)O)Cl
142.582
142.58
≥98.0% (GC)

Chemical Identifiers

615-62-3
142.582
VQZRLBWPEHFGCD-UHFFFAOYSA-N
14853
CC1=C(C=C(C=C1)O)Cl
C7H7ClO
MFCD00060319
phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol
3-chloro-4-methylphenol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0900

TSCA : Yes

Documents