missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-N,N-dimethylaniline, TCI America™

Manufacturer:  TCI America C319225G

 View more versions of this product

Catalog No. C319225G



Specifications

Specifications

3-Chloro-N,N-dimethylaniline
Undesignated
6848-13-1
MFCD00018456
CHHCCYVOJBBCIY-UHFFFAOYSA-N
3-chloro-N,N-dimethylaniline
23285
Clear Liquid at 20°C
234°C
25g
C8H10ClN
n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride
CN(C)C1=CC(=CC=C1)Cl
155.625
155.63
≥97.0% (GC,T)

Chemical Identifiers

6848-13-1
155.625
CHHCCYVOJBBCIY-UHFFFAOYSA-N
23285
CN(C)C1=CC(=CC=C1)Cl
C8H10ClN
MFCD00018456
n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride
3-chloro-N,N-dimethylaniline
Safety and Handling

Safety and Handling

TSCA : Yes

Documents