missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-N-methylaniline, 97%

Manufacturer:  Alfa Aesar™ A1241806

 View more versions of this product

Catalog No. AAA1241806


 

Specifications

Specifications

3-Chloro-N-methylaniline
243°C to 244°C
7006-52-2
C7H8ClN
UN2810
N-Methyl-3-chloroaniline
CNC1=CC(=CC=C1)Cl
141.598
141.6
>110°C (230°F)
5g
97%
MFCD00052014
2802319
WFGYSQDPURFIFL-UHFFFAOYSA-N
3-chloro-N-methylaniline
138900

Chemical Identifiers

7006-52-2
141.598
WFGYSQDPURFIFL-UHFFFAOYSA-N
138900
C7H8ClN
MFCD00052014
N-Methyl-3-chloroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents