missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-N-(4-methoxyphenyl)benzamide, 97%

Manufacturer:  Alfa Aesar™ H5630503

 View more versions of this product

Catalog No. AAH5630503



Specifications

Specifications

3-Chloro-N-(4-methoxyphenyl)benzamide
158525-81-6
MFCD01033999
OASYJKCQPDJQQR-UHFFFAOYSA-N
3-chloro-N-(4-methoxyphenyl)benzamide
835440
97%
1g
C14H12ClNO2
3-chloro-n-4-methoxyphenyl benzamide, cbmicro_012214, 3-chloro-n~1~-4-methoxyphenyl benzamide, 3-chlorophenyl-n-4-methoxyphenyl carboxamide
COC1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)Cl
261.705
261.71

Chemical Identifiers

158525-81-6
261.705
OASYJKCQPDJQQR-UHFFFAOYSA-N
835440
COC1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)Cl
C14H12ClNO2
MFCD01033999
3-chloro-n-4-methoxyphenyl benzamide, cbmicro_012214, 3-chloro-n~1~-4-methoxyphenyl benzamide, 3-chlorophenyl-n-4-methoxyphenyl carboxamide
3-chloro-N-(4-methoxyphenyl)benzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents