Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-L-tyrosine, 97%

$43.41 - $99.74

Chemical Identifiers

Chemical Name or Material 3-Chloro-L-tyrosine
CAS 7423-93-0
Molecular Formula C9H10ClNO3
Formula Weight 215.64
MDL Number MFCD00045865
Beilstein 2941263
Synonym 3-Chloro-4-hydroxy-L-phenylalanine; H-Tyr(3-Cl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0767503

  SDS
alfa aesar™
L0767503
1g Each for $99.74

AAL07675MD

  SDS
alfa aesar™
L07675MD
250mg Each for $43.41
Description & Specifications

Specifications

Formula Weight 215.64
Chemical Name or Material 3-Chloro-L-tyrosine
Molecular Formula C9H10ClNO3
MDL Number MFCD00045865
CAS 7423-93-0
Beilstein 2941263
Synonym 3-Chloro-4-hydroxy-L-phenylalanine; H-Tyr(3-Cl)-OH
Melting Point ≈250°C
Assay 97%
Quantity 1g