Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-D-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material 3-Chloro-D-phenylalanine
CAS 80126-52-9
Formula Weight 199.63
MDL Number MFCD01631984
Synonym D-Phe(3-Cl)-OH; (R)-2-Amino-3-(3-chlorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6323903

  SDS
alfa aesar™
H6323903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6323906

  SDS
alfa aesar™
H6323906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63239MD

  SDS
alfa aesar™
H63239MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.63
Assay 98%
Storage Note 1 2° to 8°C
Chemical Name or Material 3-Chloro-D-phenylalanine
Synonym D-Phe(3-Cl)-OH; (R)-2-Amino-3-(3-chlorophenyl)propionic acid
MDL Number MFCD01631984
CAS 80126-52-9
Color White
Physical Form Powder
Quantity 1g
Safety and Handling

2° to 8°C