missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-D-phenylalanine, 98%

Manufacturer:  Alfa Aesar™ H63239MD

 View more versions of this product

Catalog No. AAH63239MD



Specifications

Specifications

3-Chloro-D-phenylalanine
C9H10ClNO2
3-chloro-d-phenylalanine, h-d-phe 3-cl-oh, r-2-amino-3-3-chlorophenyl propanoic acid, d-3-chlorophenylalanine, 2r-2-amino-3-3-chlorophenyl propanoic acid, d-phe 3-cl-oh, d-3-chlorophe, 3-chloro-d-phe-oh, d-phenylalanine, 3-chloro, pubchem11977
C1=CC(=CC(=C1)Cl)CC(C(=O)O)N
199.634
199.63
98%
250mg
80126-52-9
MFCD01631984
JJDJLFDGCUYZMN-MRVPVSSYSA-N
(2R)-2-amino-3-(3-chlorophenyl)propanoic acid
7009409
Powder
White

Chemical Identifiers

80126-52-9
199.634
JJDJLFDGCUYZMN-MRVPVSSYSA-N
7009409
C1=CC(=CC(=C1)Cl)CC(C(=O)O)N
C9H10ClNO2
MFCD01631984
3-chloro-d-phenylalanine, h-d-phe 3-cl-oh, r-2-amino-3-3-chlorophenyl propanoic acid, d-3-chlorophenylalanine, 2r-2-amino-3-3-chlorophenyl propanoic acid, d-phe 3-cl-oh, d-3-chlorophe, 3-chloro-d-phe-oh, d-phenylalanine, 3-chloro, pubchem11977
(2R)-2-amino-3-(3-chlorophenyl)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents