Learn More
3-Chloro-alpha-toluenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2407314
Description
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications
3-Chloro-alpha-toluenesulfonyl chloride | |
24974-73-0 | |
C7H6Cl2O2S | |
UN3261 | |
LXTGNVLBPVVMSL-UHFFFAOYSA-N | |
(3-chlorophenyl)methanesulfonyl chloride | |
2757802 | |
96% |
69°C to 75°C | |
25 g | |
MFCD02683111 | |
3-chlorophenyl methanesulfonyl chloride, 3-chloro-phenyl-methanesulfonyl chloride, chloro 3-chlorophenyl methyl sulfone, benzenemethanesulfonyl chloride, 3-chloro, pubchem5501, acmc-20an5u, 3-chlorobenzylsulfonyl chloride, 3-chlorophenyl methanesulfonylchloride, benzenemethanesulfonylchloride,3-chloro, benzenemethanesulfonylchloride, 3-chloro | |
C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl | |
225.083 | |
225.09 |
Chemical Identifiers
24974-73-0 | |
225.083 | |
LXTGNVLBPVVMSL-UHFFFAOYSA-N | |
2757802 | |
C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl |
C7H6Cl2O2S | |
MFCD02683111 | |
3-chlorophenyl methanesulfonyl chloride, 3-chloro-phenyl-methanesulfonyl chloride, chloro 3-chlorophenyl methyl sulfone, benzenemethanesulfonyl chloride, 3-chloro, pubchem5501, acmc-20an5u, 3-chlorobenzylsulfonyl chloride, 3-chlorophenyl methanesulfonylchloride, benzenemethanesulfonylchloride,3-chloro, benzenemethanesulfonylchloride, 3-chloro | |
(3-chlorophenyl)methanesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only