Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-6-methylpyridazine, 97%

$177.01 - $684.97

Chemical Identifiers

Chemical Name or Material 3-Chloro-6-methylpyridazine
CAS 1121-79-5
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD00052905
Beilstein 108655
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1348006

 
alfa aesar™
A1348006
5g Each for $177.01

AAA1348014

 
alfa aesar™
A1348014
25g Each for $684.97
Description & Specifications

Specifications

Formula Weight 128.56
Chemical Name or Material 3-Chloro-6-methylpyridazine
Assay 97%
Molecular Formula C5H5ClN2
MDL Number MFCD00052905
CAS 1121-79-5
Beilstein 108655
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 55° to 59°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.