missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-6-iodopyridazine, 97%

Manufacturer:  Alfa Aesar™ H6387906

 View more versions of this product

Catalog No. AAH6387906



Description

Description

Applications
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
Specifications

Specifications

3-Chloro-6-iodopyridazine
5g
C4H2ClIN2
3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine
C1=CC(=NN=C1Cl)I
240.428
240.43
97%
110°C to 120°C
135034-10-5
MFCD08275187
PNEPCDPKMXJYIQ-UHFFFAOYSA-N
3-chloro-6-iodopyridazine
15418839
Solid

Chemical Identifiers

135034-10-5
240.428
PNEPCDPKMXJYIQ-UHFFFAOYSA-N
15418839
C1=CC(=NN=C1Cl)I
C4H2ClIN2
MFCD08275187
3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine
3-chloro-6-iodopyridazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents