Please login to your online account to display your discounted pricing

3-Chloro-5-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-(trifluoromethyl)phenylboronic acid
Assay Percent Range 95%
Molecular Formula C7H5BClF3O2
Formula Weight 224.38
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC445370010

  SDS
thermo fisher scientific
AC445370010
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445370050

  SDS
thermo fisher scientific
AC445370050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.38
Quantity 5g
CAS 1160561-31-8
Molecular Formula C7H5BClF3O2
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)phenylboronic acid
Assay Percent Range 95%