Please login to your online account to display your discounted pricing

3-Chloro-5-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC445370050

 View more versions of this product

Catalog No. AC445370050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 224.38
Quantity 5g
CAS 1160561-31-8
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)phenylboronic acid
Molecular Formula C7H5BClF3O2
Assay Percent Range 95%

Specifications

Chemical Name or Material 3-Chloro-5-(trifluoromethyl)phenylboronic acid
Assay Percent Range 95%
Molecular Formula C7H5BClF3O2
Formula Weight 224.38