Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-(trifluoromethyl)benzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzyl bromide
CAS 886496-91-9
Molecular Formula C8H5BrClF3
Formula Weight 273.48
MDL Number MFCD04115849
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670903

 
alfa aesar™
H2670903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2670906

 
alfa aesar™
H2670906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.48
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzyl bromide
Assay 97%
Molecular Formula C8H5BrClF3
MDL Number MFCD04115849
CAS 886496-91-9
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
API. As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Used frequently in organic synthesis. Find application as propellants.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.