Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-(trifluoromethyl)benzoic acid, 97%

$109.24 - $599.69

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzoic acid
CAS 53985-49-2
Molecular Formula C8H4ClF3O2
Formula Weight 224.57
MDL Number MFCD04115846
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2639903

  SDS
alfa aesar™
H2639903
1g Each for $109.24

AAH2639909

  SDS
alfa aesar™
H2639909
10g Each for $599.69
Description & Specifications

Specifications

Formula Weight 224.57
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzoic acid
Molecular Formula C8H4ClF3O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 53985-49-2
Melting Point 98° to 101°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD04115846
Assay 97%
Health Hazard 1 Warning
Quantity 1g

Applications
2-Chloro-5-(trifluoromethyl)benzoic acid may be used as surrogate standard to investigate the exposure and home contamination in urine and hand wipe samples collected from farmers.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.