Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 97%

$115.12 - $369.16

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester
CAS 942069-65-0
Molecular Formula C13H15BClF3O2
Formula Weight 306.52
MDL Number MFCD11856039
Synonym 2-[3-Chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5171203

  SDS
alfa aesar™
H5171203
1g Each for $369.16

AAH51712MD

  SDS
alfa aesar™
H51712MD
250mg Each for $115.12
Description & Specifications

Specifications

Formula Weight 306.52
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C13H15BClF3O2
MDL Number MFCD11856039
CAS 942069-65-0
Synonym 2-[3-Chloro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Quantity 1g