Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-5'-(trifluoromethyl)acetophenone, 97%

$94.47 - $518.32

Chemical Identifiers

Chemical Name or Material 3'-Chloro-5'-(trifluoromethyl)acetophenone
CAS 886497-11-6
Molecular Formula C9H6ClF3O
Formula Weight 222.6
MDL Number MFCD04115856
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2645003

  SDS
alfa aesar™
H2645003
1g Each for $94.47

AAH2645009

  SDS
alfa aesar™
H2645009
10g Each for $518.32
Description & Specifications

Specifications

Formula Weight 222.6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Molecular Formula C9H6ClF3O
CAS 886497-11-6
Assay 97%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3'-Chloro-5'-(trifluoromethyl)acetophenone
MDL Number MFCD04115856
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.