Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-(trifluoromethoxy)phenol, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)phenol
Molecular Formula C7H4ClF3O2
Formula Weight 212.56
MDL Number MFCD09832330
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3257903

  SDS
alfa aesar™
H3257903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32579MD

  SDS
alfa aesar™
H32579MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.56
Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)phenol
Assay 97%
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H4ClF3O2
MDL Number MFCD09832330
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.