Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-(trifluoromethoxy)benzaldehyde, 97%

$158.22 - $474.64

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)benzaldehyde
CAS 433926-48-8
Molecular Formula C8H4ClF3O2
Formula Weight 224.57
MDL Number MFCD06660305
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2678803

  SDS
alfa aesar™
H2678803
1g Each for $158.22

AAH2678806

  SDS
alfa aesar™
H2678806
5g Each for $474.64
Description & Specifications

Specifications

Formula Weight 224.57
CAS 433926-48-8
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H4ClF3O2
MDL Number MFCD06660305
Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)benzaldehyde
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
3-Chloro-5-(trifluoromethoxy)benzaldehyde is used as pharmaceutical intermediates

Solubility
Soluble in water (3.3 g/L (20°C)).

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.