Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-methoxybenzeneboronic acid, 94%

$157.00 - $392.51

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-methoxybenzeneboronic acid
CAS 915201-07-9
Molecular Formula C7H8BClO3
Formula Weight 186.4
MDL Number MFCD03095046
Synonym 3-Chloro-5-methoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2744203

 
alfa aesar™
H2744203
1g Each for $392.51

AAH27442MD

 
alfa aesar™
H27442MD
250mg Each for $157.00
Description & Specifications

Specifications

Formula Weight 186.4
Chemical Name or Material 3-Chloro-5-methoxybenzeneboronic acid
Assay 94%
Molecular Formula C7H8BClO3
MDL Number MFCD03095046
CAS 915201-07-9
Synonym 3-Chloro-5-methoxyphenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 185° to 189°C
Quantity 1g

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.