Learn More
3-Chloro-5-fluoropyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3017006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Chloro-5-fluoropyridine | |
25°C to 27°C | |
C5H3ClFN | |
MFCD05663707 | |
UN1325 | |
LTILYPOSFJLPSK-UHFFFAOYSA-N | |
3-chloro-5-fluoropyridine | |
12031536 | |
98% |
514797-99-0 | |
41°C (106°F) | |
1.5015 | |
5 g | |
5-chloro-3-fluoropyridine, 3-fluoro-5-chloropyridine, 3-chloro-5-fluoro-pyridine, pyridine, 3-chloro-5-fluoro, pubchem6620, acmc-1axgq, pyridine,3-chloro-5-fluoro, 3-chloro-5-fluoropyridine, abbypharma ap-14-5727 | |
C1=C(C=NC=C1Cl)F | |
131.534 | |
131.54 |
Chemical Identifiers
514797-99-0 | |
131.534 | |
LTILYPOSFJLPSK-UHFFFAOYSA-N | |
12031536 | |
C1=C(C=NC=C1Cl)F |
C5H3ClFN | |
MFCD05663707 | |
5-chloro-3-fluoropyridine, 3-fluoro-5-chloropyridine, 3-chloro-5-fluoro-pyridine, pyridine, 3-chloro-5-fluoro, pubchem6620, acmc-1axgq, pyridine,3-chloro-5-fluoro, 3-chloro-5-fluoropyridine, abbypharma ap-14-5727 | |
3-chloro-5-fluoropyridine |
Safety and Handling
GHS H Statement
H318-H228-H302-H312-H332-H335-H315
Causes serious eye damage.
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H312+H332-H315-H318-H335
DOTInformation : Hazard Class: 4.1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only