Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluorophenylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluorophenylacetonitrile
CAS 493038-93-0
Molecular Formula C8H5ClFN
Formula Weight 169.59
MDL Number MFCD04038259
UN Number UN3276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3041003

  SDS
alfa aesar™
H3041003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3041006

  SDS
alfa aesar™
H3041006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.59
CAS 493038-93-0
UN Number UN3276
Quantity 1g
Health Hazard 2 GHS H Statement
H300-H311-H331-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Chemical Name or Material 3-Chloro-5-fluorophenylacetonitrile
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD04038259
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Molecular Formula C8H5ClFN
Health Hazard 1 Danger

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H300-H311-H331-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.