Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluorobenzylamine hydrochloride

$640.34 - $640.34

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluorobenzylamine hydrochloride
Molecular Formula C7H7ClFN·HCl
Formula Weight 196.05
MDL Number MFCD11113045
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5113103

  SDS
alfa aesar™
H5113103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5113106

  SDS
alfa aesar™
H5113106
5g Each for $640.34
Description & Specifications

Specifications

Formula Weight 196.05
Chemical Name or Material 3-Chloro-5-fluorobenzylamine hydrochloride
Molecular Formula C7H7ClFN·HCl
MDL Number MFCD11113045
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.