Please login to your online account to display your discounted pricing

3-chloro-5-fluorobenzyl Bromide, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374700010

 View more versions of this product

Catalog No. AC374700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Low Melting Crystals, Crystalline Mass or Liquid After Melting
Formula Weight 223.47
Quantity 1g
Color White to off-white
Melting Point 25° to 30°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Linear Formula ClC6H3(F)CH2Br
Infrared Spectrum Authentic
Molecular Formula C7H5BrClF
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 493024-39-8
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 3-Chloro-5-fluorobenzyl bromide
MDL Number MFCD03788472
Assay Percent Range 97%

Specifications

Chemical Name or Material 3-Chloro-5-fluorobenzyl bromide
CAS 493024-39-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H5BrClF
Linear Formula ClC6H3(F)CH2Br
Formula Weight 223.47
MDL Number MFCD03788472
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents