Learn More
3-chloro-5-fluorobenzyl Bromide, 97%, Thermo Scientific™
$457.77 - $457.77
Chemical Identifiers
CAS | 493024-39-8 |
---|---|
Molecular Formula | C7H5BrClF |
Molecular Weight (g/mol) | 223.47 |
MDL Number | MFCD03788472 |
InChI Key | PKSAHOPFPPAUOD-UHFFFAOYSA-N |
Synonym | 3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene |
PubChem CID | 2734834 |
IUPAC Name | 1-(bromomethyl)-3-chloro-5-fluorobenzene |
SMILES | FC1=CC(Cl)=CC(CBr)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC374700050
|
Thermo Scientific Chemicals
374700050 |
5 g | Glass bottle |
N/A
|
|
||||
Chemical Identifiers
493024-39-8 | |
223.47 | |
PKSAHOPFPPAUOD-UHFFFAOYSA-N | |
2734834 | |
FC1=CC(Cl)=CC(CBr)=C1 |
C7H5BrClF | |
MFCD03788472 | |
3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene | |
1-(bromomethyl)-3-chloro-5-fluorobenzene |
Specifications
493024-39-8 | |
100.0 | |
White | |
96% min. (GC) | |
C7H5BrClF | |
MFCD03788472 | |
3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene | |
FC1=CC(Cl)=CC(CBr)=C1 | |
223.47 | |
223.47 | |
Low Melting Crystals, Crystalline Mass or Liquid After Melting |
96.0 | |
25°C to 30°C | |
Authentic | |
Glass bottle | |
ClC6H3(F)CH2Br | |
5 g | |
PKSAHOPFPPAUOD-UHFFFAOYSA-N | |
1-(bromomethyl)-3-chloro-5-fluorobenzene | |
2734834 | |
97% | |
3-Chloro-5-fluorobenzyl bromide |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only