Please login to your online account to display your discounted pricing

3-Chloro-5-fluorobenzoic acid, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374760050

 View more versions of this product

Catalog No. AC374760050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 174.56
Physical Form Crystalline Powder and Granules
Quantity 5g
Color Off-white to beige
CAS Max % 100.0
CAS Min % 96.0
MDL Number MFCD00973904
Linear Formula FC6H3(Cl)COOH
CAS 25026-64-6
Molecular Formula C7H4ClFO2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 3-Chloro-5-fluorobenzoic acid

Specifications

Chemical Name or Material 3-Chloro-5-fluorobenzoic acid
CAS 25026-64-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4ClFO2
Linear Formula FC6H3(Cl)COOH
Formula Weight 174.56
MDL Number MFCD00973904
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents