Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-5-fluorobenzoic acid, 97%

$107.30 - $588.00

Chemical Identifiers

Chemical Name or Material 3-Chloro-5-fluorobenzoic acid
CAS 25026-64-6
Molecular Formula C7H4ClFO2
Formula Weight 174.56
MDL Number MFCD00973904
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2656703

  SDS
alfa aesar™
H2656703
1g Each for $107.30

AAH2656709

  SDS
alfa aesar™
H2656709
10g Each for $588.00
Description & Specifications

Specifications

Formula Weight 174.56
Chemical Name or Material 3-Chloro-5-fluorobenzoic acid
Assay 97%
Molecular Formula C7H4ClFO2
MDL Number MFCD00973904
CAS 25026-64-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
3-Chloro-5-fluorobenzoic acid is used as a pharmaceutical intermediate. It acts as a protective reagent and is used in chemical research.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.