Please login to your online account to display your discounted pricing

3-chloro-5-fluorobenzaldehyde, 97%, ACROS Organics™


Quantity 20g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374750200

 View more versions of this product

Catalog No. AC374750200

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 158.56
Physical Form Low Melting Solid or Liquid
Quantity 20g
Color White to yellow or clear
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Linear Formula FC6H3(Cl)CHO
Infrared Spectrum Authentic
Molecular Formula C7H4ClFO
Chemical Name or Material 3-chloro-5-fluorobenzaldehyde
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
CAS 90390-49-1
Assay Percent Range 97%
MDL Number MFCD02261734

Specifications

Chemical Name or Material 3-chloro-5-fluorobenzaldehyde
CAS 90390-49-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4ClFO
Linear Formula FC6H3(Cl)CHO
Formula Weight 158.56
MDL Number MFCD02261734
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents